Invitrogen
Manufacturer Code:PA579680
Catalog # PIPA579680
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived human MMP16 recombit protein (Position: Y120-I296). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: matrix metallopeptidase 16 (membrane-inserted); matrix metalloproteinase 16; Matrix metalloproteinase-16; Matrix metalloproteinase-16 MMP-16 MMP-X2 Membrane-type matrix metalloproteinase 3 MT-MMP 3 MTMMP3 Membrane-type-3 matrix metalloproteinase MT3-MMP MT3MMP MMP16 MMPX2; Membrane type 3-MMP; Membrane-type matrix metalloproteinase 3; Membrane-type-3 matrix metalloproteinase; MMP-16; MMP-X2; MT-MMP 3; MT3-MMP; MT3MMP; MTMMP3; Putative transmembrane protein C8orf57
Gene Aliases: C8orf57; MMP-X2; MMP16; MMPX2; MT-MMP2; MT-MMP3; MT3-MMP; Mt3mmp
UniProt ID: (Human) P51512, (Rat) O35548, (Mouse) Q9WTR0
Entrez Gene ID: (Human) 4325, (Rat) 65205, (Mouse) 17389
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.