Invitrogen
Manufacturer Code:MA535044
Catalog # PIMA535044
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0211 |
Immunogen | A synthesized peptide derived from human MT1-MMP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: AI325305 cb386 EC 3.4.24.80 matrix metallo protease Matrix metalloproteinase-14 Membrane-type matrix metalloproteinase 1 Membrane-type-1 matrix metalloproteinase MMP MMP14 MMP-14 MMPs MMP-X MT-MMP MMP-X1 MT1MMP MT1-MMP Mtmmp MT-MMP MT-MMP 1 MT-MMP MT1-MMP MTMMP1 MT-MMP-1 sabe sb:cb386 WNCHRS wu:fb72b09; matrix metallopeptidase 14 (membrane-inserted); matrix metalloproteinase 14 (membrane-inserted); matrix metalloproteinase 14 membrane-inserted; matrix metalloproteinase 14, membrane-inserted; Matrix metalloproteinase-14; membrane type 1 metalloprotease; membrane type 1-matrix metalloproteinase; Membrane type 1-MMP; Membrane-type matrix metalloproteinase 1; Membrane-type-1 matrix metalloproteinase; MMP-14; MMP-X1; MT-MMP; MT-MMP 1; MT1-MMP; MT1MMP; MTMMP1; type 1 matrix metalloprotease 14
Gene Aliases: AI325305; MMP-14; MMP-X1; MMP14; MT-MMP; MT-MMP 1; MT-MMP-1; MT1-MMP; MT1MMP; Mtmmp; MTMMP1; sabe; WNCHRS
UniProt ID: (Human) P50281, (Mouse) P53690, (Rat) Q10739
Entrez Gene ID: (Human) 4323, (Mouse) 17387, (Rat) 81707
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.