OriGene
Manufacturer Code:CF506806
Catalog # 50-166-4454
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2D8 |
Immunogen | Human recombit protein fragment corresponding to amino acids 104-471 of human MMP13 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CLG3 MANDP1 MDST MMP-13; Collagenase 3; matrix metalloproteinase 13 (collagenase 3); Matrix metalloproteinase-13; MMP-13
Gene Aliases: CLG3; MANDP1; MMP-13; MMP13
UniProt ID: (Human) P45452
Entrez Gene ID: (Human) 4322
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.