Invitrogen
Manufacturer Code:PA586640
Catalog # PIPA586640
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 400-450 of Human MMP-12. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Macrophage elastase; Macrophage metalloelastase; Macrophage metalloelastase MME 3.4.24.65 Macrophage elastase ME hME Matrix metalloproteinase-12 MMP-12 MMP12 HME; macrophage-metalloelastase; matrix metallopeptidase 12 (macrophage elastase); matrix metalloproteinase 12; matrix metalloproteinase 12 (macrophage elastase); Matrix metalloproteinase-12; ME; MME; MMP-12
Gene Aliases: AV378681; HME; ME; MME; Mmel; MMP-12; MMP12
UniProt ID: (Human) P39900, (Mouse) P34960, (Rat) Q63341
Entrez Gene ID: (Human) 4321, (Mouse) 17381, (Rat) 117033
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.