Proteintech
Manufacturer Code:229891AP150UL
Catalog # 50-173-3976
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MMP12 Fusion Protein Ag19068 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HME Macrophage elastase Macrophage metalloelastase Matrix metalloproteinase 12 ME MME MMP 12 MMP12; Macrophage elastase; Macrophage metalloelastase; macrophage-metalloelastase; matrix metallopeptidase 12 (macrophage elastase); matrix metalloproteinase 12; matrix metalloproteinase 12 (macrophage elastase); Matrix metalloproteinase-12; ME; MME; MMP-12
Gene Aliases: AV378681; HME; ME; MME; Mmel; MMP-12; MMP12
UniProt ID: (Human) P39900, (Mouse) P34960
Entrez Gene ID: (Human) 4321, (Mouse) 17381
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.