Abnova
Manufacturer Code:MAB20194
Catalog # 89-336-594
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | CDG-13 |
Immunogen | A synthetic peptide corresponding to human MMP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C for one year. After reconstitution at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. |
Rabbit Monoclonal Antibody.
Protein Aliases: 22 kDa interstitial collagenase; 27 kDa interstitial collagenase; CLG CLGN; Fibroblast collagenase; Interstitial collagenase; matrix metallopeptidase 1 (interstitial collagenase); matrix metalloprotease 1; matrix metalloproteinase 1; Matrix metalloproteinase-1; MMP-1
Gene Aliases: CLG; CLGN; MMP1
UniProt ID: (Human) P03956
Entrez Gene ID: (Human) 4312
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.