Invitrogen
Manufacturer Code:MA537734
Catalog # PIMA537734
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1054CT20.4.3 |
Immunogen | recombinant protein from human MMP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 22 kDa interstitial collagenase; 22 kDa interstitial collagenase 27 kDa interstitial collagenase CLG CLGN Collagenase collagenase-like A fibroblast collagenase Interstitial collagenase interstitial collagenase A matrix metallo protease matrix metallopeptidase 1 matrix metallopeptidase 1 (interstitial collagenase) matrix metallopeptidase 1a matrix metallopeptidase 1a (interstitial collagenase) matrix metalloprotease 1 matrix metalloproteinase 1 matrix metalloproteinase 1a (interstitial collagenase) Matrix metalloproteinase1 Matrix metalloproteinase-1 Matrix metalloproteinase-1a Mcola Mcol-A MMP MMP1 MMP-1 Mmp1a MMP-1a MMPs; 27 kDa interstitial collagenase; Fibroblast collagenase; Interstitial collagenase; matrix metallopeptidase 1 (interstitial collagenase); matrix metalloprotease 1; matrix metalloproteinase 1; Matrix metalloproteinase-1; MMP-1
Gene Aliases: CLG; CLGN; MMP1
UniProt ID: (Human) P03956
Entrez Gene ID: (Human) 4312
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.