R&D Systems
Manufacturer Code:AF513SP
Catalog # AF513SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human MMP-3 Tyr18-Cys477 (Lys45Glu) Accession # P08254 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CHDS6 EC 3.4.24 EC 3.4.24.17 matrix metallopeptidase 3 (stromelysin 1 progelatinase) matrix metalloproteinase 3 (stromelysin 1 progelatinase) Matrix metalloproteinase-3 MGC126102 MGC126103 MMP3 MMP-3 proteoglycanase SL-1 STMY STMY1MGC126104 STR1 Stromelysin 1 stromelysin-1 Transin-1; matrix metalloproteinase 3 (stromelysin 1, progelatinase); Matrix metalloproteinase-3; MMP-3; proteoglycanase; SL-1; Stromelysin-1; Transin-1
Gene Aliases: CHDS6; MMP-3; MMP3; SL-1; STMY; STMY1; STR1
UniProt ID: (Human) P08254
Entrez Gene ID: (Human) 4314
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.