Novus Biologicals
Manufacturer Code:NB200193SS
Catalog # NB200193SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Human C-terminal peptide (aa. 475-490). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 72 kDa gelatinase; 72 kDa gelatinase CLG4 CLG4A72 kDa type IV collagenase collagenase type IV-A EC 3.4.24 EC 3.4.24.24 Gelatinase A matrix metallopeptidase 2 (gelatinase A 72kDa gelatinase 72kDa type IVcollagenase) matrix metalloproteinase 2 (gelatinase A 72kD gelatinase 72kD type IVcollagenase) Matrix metalloproteinase-2 matrix metalloproteinase-II MMP-2 MMP-2 ihc MMP-2 mouse MMP-2 western blot MMP-II MONA neutrophil gelatinase TBE-1matrix metalloproteinase 2 (gelatinase A 72kDa gelatinase 72kDa type IVcollagenase); 72 kDa type IV collagenase; collagenase type IV-A; Gelatinase A; matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IV collagenase); Matrix metalloproteinase-2; matrix metalloproteinase-II; MMP-2; neutrophil gelatinase; PEX; TBE-1
Gene Aliases: CLG4; CLG4A; MMP-2; MMP-II; MMP2; MONA; TBE-1
UniProt ID: (Human) P08253
Entrez Gene ID: (Human) 4313
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.