R&D Systems
Manufacturer Code:FAB9181N
Catalog # FAB9181N
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 128527 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human MMP-14/MT1-MMP Tyr112-Ala541 (predicted) Accession # P50281 |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 °C as supplied. |
For Research Use Only
Protein Aliases: EC 3.4.24 EC 3.4.24.80 matrix metallopeptidase 14 (membrane-inserted) matrix metalloproteinase 14 (membrane-inserted) matrix metalloproteinase-14 membrane type 1 metalloprotease Membrane-type matrix metalloproteinase 1 Membrane-type-1 matrix metalloproteinase MMP14 MMP-14 MMP-X1 MT1MMP MT1-MMP MT1-MMPMTMMP1 MT-MMP 1 MT-MMP1; matrix metallopeptidase 14 (membrane-inserted); Matrix metalloproteinase-14; membrane type 1 metalloprotease; Membrane-type matrix metalloproteinase 1; Membrane-type-1 matrix metalloproteinase; MMP-14; MMP-X1; MT-MMP 1; MT1-MMP
Gene Aliases: MMP-14; MMP-X1; MMP14; MT-MMP; MT-MMP 1; MT1-MMP; MT1MMP; MTMMP1; WNCHRS
UniProt ID: (Human) P50281
Entrez Gene ID: (Human) 4323
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.