Novus Biologicals
Manufacturer Code:DDX0281HRPO0.05MG
Catalog # DDX0281HRP5
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 706F9.01 |
Immunogen | MMP-12/HME was used as immunogen. |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C in the dark. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 3.4.24 hME HMEEC 3.4.24.65 Macrophage elastase macrophage metalloelastase matrix metallopeptidase 12 (macrophage elastase) matrix metalloproteinase 12 (macrophage elastase) Matrix metalloproteinase-12 ME MGC138506 MME MMP-12; Macrophage elastase; Macrophage metalloelastase; matrix metallopeptidase 12 (macrophage elastase); matrix metalloproteinase 12 (macrophage elastase); Matrix metalloproteinase-12; ME; MME; MMP-12
Gene Aliases: HME; ME; MME; MMP-12; MMP12
UniProt ID: (Human) P39900
Entrez Gene ID: (Human) 4321
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.