R&D Systems
Manufacturer Code:AF3467
Catalog # AF3467
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse MMP-12 Ala18-Cys462 Accession # EDL24933 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.4.24 hME HMEEC 3.4.24.65 Macrophage elastase macrophage metalloelastase matrix metallopeptidase 12 (macrophage elastase) matrix metalloproteinase 12 (macrophage elastase) Matrix metalloproteinase-12 ME MGC138506 MME MMP12 MMP-12; Macrophage elastase; Macrophage metalloelastase; matrix metallopeptidase 12 (macrophage elastase); matrix metalloproteinase 12 (macrophage elastase); Matrix metalloproteinase-12; ME; MME; MMP-12
Gene Aliases: HME; ME; MME; MMP-12; MMP12
UniProt ID: (Human) P39900
Entrez Gene ID: (Human) 4321
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.