Invitrogen
Manufacturer Code:PA577559
Catalog # PIPA577559
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)NTSLKLRKFDSLRR corresponding to amino acid residues 118-131 of rat Magnesium Transporter MMgT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ER membrane protein complex subunit 5; Magnesium Transporter MMgT1; Membrane magnesium transporter 1; membrane magnesium transporter 1 {ECO:0000250|UniProtKB:Q8K273}; mmgt1 {ECO:0000250|UniProtKB:Q8K273}; Transmembrane protein 32; transmembrane protein 32 {ECO:0000312|EMBL:AAI68927.1}
Gene Aliases: 9630048L06Rik; BC032271; Emc5; mir-5116; Mir5116; Mmgt1; RGD1566339; Tmem32
UniProt ID: (Mouse) Q8K273, (Rat) B5DF51
Entrez Gene ID: (Mouse) 236792, (Rat) 302864
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.