Proteintech
Manufacturer Code:600343IG150UL
Catalog # 50-173-6198
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4B9D9 |
Immunogen | MMECD10 Fusion Protein Ag0427 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Atriopeptidase; Atriopeptidase CALLA CD10 Enkephalinase EPN membrane metallo endopeptidase MME NEP Neprilysin Neutral endopeptidase Neutral endopeptidase 24.11; CALLA; CD10; common acute lymphoblastic leukemia antigen; Common acute lymphocytic leukemia antigen; Enkephalinase; membrane metallo endopeptidase; membrane metallo-endopeptidase (neutral endopeptidase, enkephalinase, CALLA, CD10); membrane metallo-endopeptidase variant 1; membrane metallo-endopeptidase variant 2; NEP; Neprilysin; neprilysin-390; neprilysin-411; Neutral endopeptidase 24.11; SFE; Skin fibroblast elastase
Gene Aliases: 6030454K05Rik; C85356; CALLA; CD10; CMT2T; EPN; MME; NEP; SFE
UniProt ID: (Human) Q3MIX4, (Mouse) Q61391
Entrez Gene ID: (Human) 4311, (Pig) 100511536, (Mouse) 17380
Molecular Function:
hydrolase
metalloprotease
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.