Abnova
Manufacturer Code:MAB4606
Catalog # 89-111-928
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MEM-78 |
Immunogen | Native purified MME from NALM-6 human pre-B cell line. |
Conjugate | PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store in the dark at 4°C. Do not freeze.<br />Avoid prolonged exposure to light.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against native MME.
For Research Use Only
Protein Aliases: Atriopeptidase; CALLA; CD10; Common acute lymphocytic leukemia antigen; DKFZp686O16152; Enkephalinase; membrane metallo-endopeptidase (neutral endopeptidase, enkephalinase, CALLA, CD10); membrane metallo-endopeptidase variant 1; membrane metallo-endopeptidase variant 2; MGC126681; MGC126707; NEP; Neprilysin; neprilysin-390; neprilysin-411; Neutral endopeptidase 24.11; SFE; Skin fibroblast elastase
Gene Aliases: CALLA; CD10; CMT2T; EPN; MME; NEP; SFE
UniProt ID: (Human) Q3MIX4
Entrez Gene ID: (Human) 4311
Molecular Function:
hydrolase
metalloprotease
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.