Novus Biologicals
Manufacturer Code:NBP15470820UL
Catalog # NBP15470820
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to MMADHC. The peptide sequence was selected from the middle region of MMADHC (NP_056517). Peptide sequence RAEGYWADFIDPSSGLAFFGPYTNNTLFETDERYRHLGFSVDDLGCCKVI. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CblD; cblD chromosome 2 open reading frame 25 methylmalonic aciduria (cobalamin deficiency) cblD type with homocystinuria mitochondrial protein C2orf25 mitochondrial; Cobalamin trafficking protein CblD; methylmalonic aciduria (cobalamin deficiency) cblD type, with homocystinuria; Methylmalonic aciduria and homocystinuria type D protein, mitochondrial; protein C2orf25, mitochondrial
Gene Aliases: C2orf25; cblD; CL25022; HSPC161; MMADHC; My011
UniProt ID: (Human) Q9H3L0
Entrez Gene ID: (Human) 27249
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.