Novus Biologicals
Manufacturer Code:NBP132513
Catalog # NBP132513
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human MMACHC. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alkylcobalamin:glutathione S-alkyltransferase; CblC; cblC DKFZP564I122 FLJ25671 methylmalonic aciduria (cobalamin deficiency) cblC type with homocystinuria methylmalonic aciduria and homocystinuria type C protein RP11-291L19.3; Cyanocobalamin reductase (cyanide-eliminating); Cyanocobalamin reductase / alkylcobalamin dealkylase; Methylmalonic aciduria and homocystinuria type C protein; MMACHC
Gene Aliases: cblC; MMACHC
UniProt ID: (Human) Q9Y4U1
Entrez Gene ID: (Human) 25974
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.