Proteintech
Manufacturer Code:214621AP150UL
Catalog # 50-173-3431
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MMACHC Fusion Protein Ag15001 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alkylcobalamin:glutathione S-alkyltransferase; CblC; cblC MMACHC RP11 291L19.3; Cyanocobalamin reductase (cyanide-eliminating); Cyanocobalamin reductase / alkylcobalamin dealkylase; Methylmalonic aciduria and homocystinuria type C protein; methylmalonic aciduria and homocystinuria type C protein homolog; MMACHC
Gene Aliases: 1810037K07Rik; cblC; MMACHC; RGD1310806
UniProt ID: (Human) Q9Y4U1, (Mouse) Q9CZD0
Entrez Gene ID: (Human) 25974, (Mouse) 67096, (Rat) 313520
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.