Novus Biologicals
Manufacturer Code:NBP272730V
Catalog # NBP272730V
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2G5 |
Immunogen | Full length recombinant protein of human MMAB (NP_443077) produced in HEK293T cells. |
Conjugate | DyLight 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: aquocob(I)alamin vitamin B12s adenosyltransferase; aquocob(I)alamin vitamin B12s adenosyltransferase ATP:cob(I)alamin adenosyltransferase ATP:corrinoid adenosyltransferase ATR cblB c-diamide adenosyltransferase mitochondrial cob Cob(I)alamin adenosyltransferase cob(I)yrinic acid a EC 2.5.1.17 methylmalonic aciduria (cobalamin deficiency) cblB type methylmalonic aciduria (cobalamin deficiency) type B Methylmalonic aciduria type B protein MGC20496; ATP:cob(I)alamin adenosyltransferase; ATP:corrinoid adenosyltransferase; cilia and flagella associated protein 23; Cob(II)alamin adenosyltransferase; Cob(II)yrinic acid a,c-diamide adenosyltransferase; Cobinamide/cobalamin adenosyltransferase; Corrinoid adenosyltransferase; Methylmalonic aciduria type B protein
Gene Aliases: ATR; cblB; CFAP23; cob; MMAB
UniProt ID: (Human) Q96EY8
Entrez Gene ID: (Human) 326625
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.