OriGene
Manufacturer Code:CF502179
Catalog # 50-166-1108
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2A7 |
Immunogen | Full length human recombit protein of human MMAB produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: aquocob(I)alamin vitamin B12s adenosyltransferase; ATP:cob(I)alamin adenosyltransferase; ATP:corrinoid adenosyltransferase; ATR CFAP23 cblB cob; cilia and flagella associated protein 23; Cob(II)alamin adenosyltransferase; Cob(II)yrinic acid a,c-diamide adenosyltransferase; Cobinamide/cobalamin adenosyltransferase; Corrinoid adenosyltransferase; Methylmalonic aciduria type B protein
Gene Aliases: ATR; cblB; CFAP23; cob; MMAB
UniProt ID: (Human) Q96EY8
Entrez Gene ID: (Human) 326625
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.