Abnova
Manufacturer Code:PAB12520
Catalog # 89-115-895
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 800-852 of human MLXIPL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. Do not freeze. |
Rabbit polyclonal antibody raised against synthetic peptide of MLXIPL.
For Research Use Only
Protein Aliases: bHLHd14; carbohydrate response element binding protein; Carbohydrate-responsive element-binding protein; CHREBP; Class D basic helix-loop-helix protein 14; MIO; MLX interacting protein-like; MLX interactor; MLX-interacting protein-like; MONDOB; WBSCR14; Williams Beuren syndrome chromosome region 14; Williams-Beuren syndrome chromosomal region 14 protein; Williams-Beuren syndrome chromosome region 14 protein 1; Williams-Beuren syndrome chromosome region 14 protein 2; Williams-Beuren syndrome chromosome region 14 protein 3; WS basic-helix-loop-helix leucine zipper protein; WS-bHLH
Gene Aliases: BHLHD14; CHREBP; MIO; MLXIPL; MONDOB; WBSCR14; WS-bHLH
UniProt ID: (Human) Q9BY05
Entrez Gene ID: (Human) 51085
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.