Invitrogen
Manufacturer Code:PA523734
Catalog # PIPA523734
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 72-100 amino acids from the N-terminal region of human MLXIP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bHLHe36; Class E basic helix-loop-helix protein 36; MIR MONDOA bHLHe36 MLX-interacting protein Mlx interactor MondoA class E basic helix-loop-helix protein 36 transcriptional activator MondoA; Mlx interactor; MLX-interacting protein; MLXIP; Transcriptional activator MondoA
Gene Aliases: AW228700; BHLHE36; KIAA0867; MIR; MLXIP; MONDOA
UniProt ID: (Human) Q9HAP2, (Mouse) Q2VPU4
Entrez Gene ID: (Human) 22877, (Mouse) 208104
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.