Novus Biologicals
Manufacturer Code:NBP2852970.1ML
Catalog # NB033226
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human MLXIP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bHLHe36; bHLHe36BHLHE36 Class E basic helix-loop-helix protein 36 KIAA0867MondoA MIRMLX-interacting protein MLX interacting protein MONDOAMlx interactor Transcriptional activator MondoA; Class E basic helix-loop-helix protein 36; Mlx interactor; MLX-interacting protein; MLXIP; Transcriptional activator MondoA
Gene Aliases: BHLHE36; KIAA0867; MIR; MLXIP; MONDOA
UniProt ID: (Human) Q9HAP2
Entrez Gene ID: (Human) 22877
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.