Proteintech
Manufacturer Code:136141AP150UL
Catalog # 50-557-533
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MLXIP Fusion Protein Ag4519 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bHLHe36; bHLHe36 KIAA0867 MIR MLX interacting protein MLXIP MONDOA; Class E basic helix-loop-helix protein 36; MLX interacting protein; Mlx interactor; MLX-interacting protein; MLXIP; Transcriptional activator MondoA
Gene Aliases: AW228700; BHLHE36; KIAA0867; MIR; MLXIP; MONDOA; RGD1560774
UniProt ID: (Human) Q9HAP2, (Mouse) Q2VPU4
Entrez Gene ID: (Human) 22877, (Rat) 304479, (Mouse) 208104
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.