Invitrogen
Manufacturer Code:PA543487
Catalog # PIPA543487
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human MLSTD2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: fatty acyl CoA reductase 1; Fatty acyl-CoA reductase 1; male sterility domain containing 2; Male sterility domain-containing protein 2; short chain dehydrogenase/reductase family 10E, member 1
Gene Aliases: 2600011M19Rik; 2900034E22Rik; 3732409C05Rik; AI850429; FAR1; MLSTD2; PFCRD; SDR10E1; UNQ2423/PRO4981
UniProt ID: (Human) Q8WVX9, (Mouse) Q922J9, (Rat) Q66H50
Entrez Gene ID: (Bovine) 525262, (Horse) 100071555, (Dog) 476863, (Rabbit) 100351559, (Human) 84188, (Mouse) 67420, (Rat) 293173, (Guinea pig) 100713905
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.