OriGene
Manufacturer Code:CF808433
Catalog # 50-166-8851
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI6E3 |
Immunogen | Human recombit protein fragment corresponding to amino acids 295-600 of human MLPH produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Exophilin-3; Melanophilin; SLAC2-A; Slp homolog lacking C2 domains a; Synaptotagmin-like protein 2a
Gene Aliases: MLPH; SLAC2-A; SLAC2A
UniProt ID: (Human) Q9BV36
Entrez Gene ID: (Human) 79083
Molecular Function:
G-protein modulator
actin binding motor protein
actin family cytoskeletal protein
cytoskeletal protein
enzyme modulator
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.