Invitrogen
Manufacturer Code:PA571740
Catalog # PIPA571740
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 357-386 amino acids from the C-terminal region of human MLNR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein-coupled receptor 38; G-protein coupled receptor 38; MLNR G-protein coupled receptor 38 Motilin receptor MTLR MTLR1 G protein-coupled receptor 38 GPR38; Motilin receptor
Gene Aliases: GPR38; MLNR; MTLR; MTLR1
UniProt ID: (Human) O43193
Entrez Gene ID: (Human) 2862
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.