Proteintech
Manufacturer Code:180471AP150UL
Catalog # 50-173-1958
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MLN51/CASC3 Fusion Protein Ag12694 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Barentsz; barentsz protein; Btz; BTZ CASC3 MLN 51 MLN51 Protein barentsz Protein CASC3; Cancer susceptibility candidate gene 3 protein; Cancer susceptibility candidate gene 3 protein homolog; mBtz; metastatic lymph node 51; Metastatic lymph node gene 51 protein; Metastatic lymph node gene 51 protein homolog; MLN 51; MLN 51 homolog; Protein barentsz; Protein CASC3
Gene Aliases: BTZ; CASC3; MLN51
UniProt ID: (Human) O15234, (Mouse) Q8K3W3, (Rat) Q8K3X0
Entrez Gene ID: (Human) 22794, (Mouse) 192160, (Rat) 259170
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.