Invitrogen
Manufacturer Code:PA5106260
Catalog # PIPA5106260
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human MLK3 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2610017K16Rik MAP3K11 MEKK11 MGC17114 mitogen activated protein kinase kinase kinase 11 mitogen-activated protein kinase kinase kinase 11 Mixed lineage kinase 3 Mlk3 MLK-3 protein-tyrosine kinase PTK1 PTK1 RHOE SH3 domain-containing proline-rich kinase SPRK Src-homology 3 domain-containing proline-rich kinase; MAP3K11; mitogen activated protein kinase kinase kinase 11; Mitogen-activated protein kinase kinase kinase 11; Mixed lineage kinase 3; protein-tyrosine kinase PTK1; SH3 domain-containing proline-rich kinase; Src-homology 3 domain-containing proline-rich kinase
Gene Aliases: 2610017K16Rik; MAP3K11; MEKK11; MLK-3; MLK3; PTK1; RHOE; SPRK
UniProt ID: (Human) Q16584, (Rat) Q66HA1, (Mouse) Q80XI6
Entrez Gene ID: (Human) 4296, (Rat) 309168, (Mouse) 26403
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.