Novus Biologicals
Manufacturer Code:NBP15891320UL
Catalog # NBP15891320
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to MAP3K11(mitogen-activated protein kinase kinase kinase 11) The peptide sequence was selected from the middle region of MAP3K11. Peptide sequence PVGQRSAKSPRREEEPRGGTVSPPPGTSRSAPGTPGTPRSPPLGLISRPR. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.11 EC 2.7.11.25 MEKK11 mitogen-activated protein kinase kinase kinase 11 Mixed lineage kinase 3 MLK3MLK-3 PTK1protein-tyrosine kinase PTK1 SH3 domain-containing proline-rich kinase SPRKMGC17114 Src-homology 3 domain-containing proline-rich kinase; MAP3K11; Mitogen-activated protein kinase kinase kinase 11; Mixed lineage kinase 3; protein-tyrosine kinase PTK1; SH3 domain-containing proline-rich kinase; Src-homology 3 domain-containing proline-rich kinase
Gene Aliases: MAP3K11; MEKK11; MLK-3; MLK3; PTK1; SPRK
UniProt ID: (Human) Q16584
Entrez Gene ID: (Human) 4296
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.