Invitrogen
Manufacturer Code:PA571884
Catalog # PIPA571884
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 110-138 amino acids from the N-terminal region of mouse Map3k10 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MAP3K9 MEKK9 MLK1 Mixed lineage kinase 1 PRKE1 Mixed lineage kinase 2 Mitogen-activated protein kinase kinase kinase 10 Protein kinase MST Mixed lineage kinase 2 Mitogen-activated protein kinase kinase kinase 10 Protein kinase MST; Mitogen-activated protein kinase kinase kinase 10; Mixed lineage kinase 2; MKN28 derived nonreceptor_type; MKN28 derived nonreceptor_type serine/threonine kinase; MKN28 kinase; Protein kinase MST; serine/threonine kinase
Gene Aliases: BC028668; BC046514; MAP3K10; MEKK10; MLK2; MST
UniProt ID: (Human) Q02779, (Mouse) Q66L42
Entrez Gene ID: (Human) 4294, (Mouse) 269881
Molecular Function: kinase non-receptor serine/threonine protein kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.