Novus Biologicals
Manufacturer Code:NBP187737
Catalog # NBP187737
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:LPSGFEHKITVQASPTLDKRKGSDGASPPASPSIIPRLRAIRLTPVDCGGSSSGSSSGGSGTWSRGGPPKKEELVGGKKKGRTWGPSSTLQKERVGGEERLKGLGEGSKQWSSS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.11 MEKK10 mitogen-activated protein kinase kinase kinase 10 Mixed lineage kinase 2 MKN28 kinase MLK2MKN28 derived nonreceptor_type serine/threonine kinase MSTEC 2.7.11.25 Protein kinase MST; Mitogen-activated protein kinase kinase kinase 10; Mixed lineage kinase 2; MKN28 derived nonreceptor_type serine/threonine kinase; MKN28 kinase; Protein kinase MST
Gene Aliases: MAP3K10; MEKK10; MLK2; MST
UniProt ID: (Human) Q02779
Entrez Gene ID: (Human) 4294
Molecular Function: kinase non-receptor serine/threonine protein kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.