Invitrogen
Manufacturer Code:PA515105
Catalog # PIPA515105
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 1070-1104 amino acids from the C-terminal region of human MLK1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: mitogen activated protein kinase kinase kinase 9; Mitogen-activated protein kinase kinase kinase 9; Mitogen-activated protein kinase kinase kinase 9 Mixed lineage kinase 1 MAP3K9 MLK1 PRKE1; Mixed lineage kinase 1; mixed lineage kinase 1 (tyr and ser/thr specificity); mixed-lineage kinase 1
Gene Aliases: E130314H24Rik; MAP3K9; MEKK9; MLK1; PRKE1; RGD1562149
UniProt ID: (Human) P80192, (Mouse) Q3U1V8
Entrez Gene ID: (Human) 4293, (Rat) 500690, (Mouse) 338372
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.