Invitrogen
Manufacturer Code:MA125669
Catalog # PIMA125669
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | G168-15 |
Immunogen | Recombit full length protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: COCA2 FCC2 HNPCC HNPCC2 MGC5172 hMLH1; DNA mismatch repair protein Mlh1; DNA mismatch repair protein Mlh1-like protein; mismatch repair protein 1; mutL homolog 1; mutL homolog 1, colon cancer, nonpolyposis type 2; MutL protein homolog 1
Gene Aliases: COCA2; FCC2; hMLH1; HNPCC; HNPCC2; I79_014127; MLH1
UniProt ID: (Human) P40692, (Rat) P97679
Entrez Gene ID: (Human) 4292, (Rat) 81685, (Chinese hamster) 100769879
Molecular Function: DNA binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.