Invitrogen
Manufacturer Code:PA585129
Catalog # PIPA585129
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human MLH1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: COCA2 antibody FCC2 antibody HNPCC antibody HNPCC2 antibody MGC5172 antibody hMLH1 antibody MLH1 antibody DNA mismatch repair protein Mlh1 antibody mutL protein homolog 1 antibody mutL homolog 1 colon cancer nonpolyposis type 2 (E. coli) antibody; DNA mismatch repair protein Mlh1; mutL homolog 1, colon cancer, nonpolyposis type 2; MutL protein homolog 1
Gene Aliases: COCA2; FCC2; hMLH1; HNPCC; HNPCC2; MLH1
UniProt ID: (Human) P40692
Entrez Gene ID: (Human) 4292
Molecular Function: DNA binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.