Invitrogen
Manufacturer Code:PA579675
Catalog # PIPA579675
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human MLH1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: colon cancer, nonpolyposis type 2; DNA mismatch repair protein Mlh1; DNA mismatch repair protein Mlh1 MutL protein homolog 1 MLH1 COCA2; mismatch repair protein 1; mutL homolog 1, colon cancer, nonpolyposis type 2; MutL protein homolog 1
Gene Aliases: 1110035C23Rik; AI317206; AI325952; AI561766; COCA2; FCC2; hMLH1; HNPCC; HNPCC2; MLH1
UniProt ID: (Human) P40692, (Rat) P97679, (Mouse) Q9JK91
Entrez Gene ID: (Human) 4292, (Rat) 81685, (Mouse) 17350
Molecular Function: DNA binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.