Proteintech
Manufacturer Code:131861AP150UL
Catalog # 50-172-8553
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MLF1IP Fusion Protein Ag3858 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CENP 50 CENP U CENP U(50) CENP50 CENPU centromere protein of 50 kDa Centromere protein U ICEN24 KLIP1 MLF1 interacting protein MLF1IP PBIP1 Polo box interacting protein 1; CENP-50; CENP-U; Centromere protein of 50 kDa; Centromere protein U; Interphase centromere complex protein 24; KSHV latent nuclear antigen interacting protein 1; KSHV latent nuclear antigen-interacting protein 1; MLF1 interacting protein; MLF1-interacting protein; myeloid leukemia factor 1 interacting protein; Polo-box-interacting protein 1
Gene Aliases: 1700029A22Rik; CENP50; CENPU; CENPU50; ICEN24; KLIP1; MLF1IP; PBIP1
UniProt ID: (Human) Q71F23, (Rat) Q4V8G7, (Mouse) Q8C4M7
Entrez Gene ID: (Human) 79682, (Rat) 306464, (Mouse) 71876
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.