Invitrogen
Manufacturer Code:PA575533
Catalog # PIPA575533
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 194-239 of Human MLF1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Hematopoietic lineage switch 7; Myelodysplasia-myeloid leukemia factor 1; Myeloid leukemia factor 1; Myeloid leukemia factor 1 Myelodysplasia-myeloid leukemia factor 1 MLF-1; testis tissue sperm-binding protein Li 49e
Gene Aliases: HLS7; MLF1
UniProt ID: (Human) P58340, (Mouse) Q9QWV4
Entrez Gene ID: (Human) 4291, (Rat) 310483, (Mouse) 17349
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.