Invitrogen
Manufacturer Code:PA515176
Catalog # PIPA515176
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 40-69 amino acids from the N-terminal region of human MYLK3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cardiac MLCK; Cardiac-MLCK; cardiac-MyBP-C associated Ca/CaM kinase; Cardiac-MyBP-C-associated Ca/CaM kinase; cardiac-specific myosin light chain kinase; MLC kinase; Myosin light chain kinase; Myosin light chain kinase 3; putative myosin light chain kinase 3
Gene Aliases: caMLCK; D830007F02Rik; MLCK; MLCK2; MYLK3
UniProt ID: (Human) Q32MK0, (Mouse) Q3UIZ8
Entrez Gene ID: (Human) 91807, (Mouse) 213435
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.