Invitrogen
Manufacturer Code:PA541042
Catalog # PIPA541042
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human MLC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: megalencephalic leukoencephalopathy with subcortical cysts 1; megalencephalic leukoencephalopathy with subcortical cysts 1 gene product; megalencephalic leukoencephalopathy with subcortical cysts 1 homolog; Megalencephalic leukoencephalopathy with subcortical cysts 1 KIAA0027 LVM MLC VL; Membrane protein MLC1
Gene Aliases: AW048630; BB074274; KIAA0027; Kiaa0027-hp; LVM; mKIAA0027; MLC; MLC1; VL; WKL1
UniProt ID: (Human) Q15049, (Mouse) Q8VHK5
Entrez Gene ID: (Dog) 607105, (Bovine) 618918, (Rabbit) 100350374, (Human) 23209, (Guinea pig) 100721749, (Mouse) 170790, (Rat) 315215
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.