Novus Biologicals
Manufacturer Code:NBP226196
Catalog # NBP226196
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-SDSANILDEVPFPAR corresponding to internal region according to NP_055981.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: LVM megalencephalic leukoencephalopathy with subcortical cysts 1 WKL1; Membrane protein MLC1
Gene Aliases: KIAA0027; LVM; MLC; MLC1; VL; WKL1
UniProt ID: (Human) Q15049
Entrez Gene ID: (Human) 23209
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.