Proteintech
Manufacturer Code:162061AP150UL
Catalog # 50-173-0790
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MKS1 Fusion Protein Ag9177 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BBS13 Meckel syndrome type 1 protein Meckel syndrome type 1 MES MKS MKS1; Meckel syndrome type 1 protein; Meckel syndrome type 1 protein homolog; POC12 centriolar protein homolog
Gene Aliases: AK190930; B8d3; BBS13; MES; MKS; MKS1; POC12
UniProt ID: (Human) Q9NXB0, (Mouse) Q5SW45, (Rat) Q499Q5
Entrez Gene ID: (Human) 54903, (Mouse) 380718, (Rat) 287612
Molecular Function: extracellular matrix protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.