Invitrogen
Manufacturer Code:PA515549
Catalog # PIPA515549
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 331-361 amino acids from the C-terminal region of human DUSP4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dual specificity protein phosphatase 4; Dual specificity protein phosphatase hVH2; MAP kinase phosphatase 2; Mitogen-activated protein kinase phosphatase 2; Mitogen-activated protein kinase phosphatase 2 MAP kinase phosphatase 2 MKP-2 Dual specificity protein phosphatase hVH2; serine/threonine specific protein phosphatase; VH1 homologous phosphatase 2
Gene Aliases: 2700078F24Rik; AI844617; BB104621; DUSP4; E130306H24Rik; HVH2; MKP-2; MKP2; TYP; VH2
UniProt ID: (Human) Q13115, (Mouse) Q8BFV3
Entrez Gene ID: (Human) 1846, (Mouse) 319520
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.