Invitrogen
Manufacturer Code:PA544322
Catalog # PIPA544322
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human MKNK2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein-coupled receptor kinase 7; MAP kinase interacting serine/threonine kinase 2; MAP kinase interacting serine/threonine kinase 2 GPRK7 MNK2; MAP kinase signal-integrating kinase 2; MAP kinase-interacting serine/threonine-protein kinase 2; MAPK signal-integrating kinase 2; mnk2
Gene Aliases: 2010016G11Rik; GPRK7; MKNK2; MNK2
UniProt ID: (Human) Q9HBH9, (Mouse) Q8CDB0, (Rat) Q5U2N4
Entrez Gene ID: (Bovine) 538519, (Human) 2872, (Dog) 612212, (Horse) 100146117, (Mouse) 17347, (Rat) 299618, (Guinea pig) 100734192
Molecular Function: cytoskeletal protein kinase microtubule family cytoskeletal protein non-motor microtubule binding protein non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.