Abnova
Manufacturer Code:H00002872A01
Catalog # 89-104-574
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 1 a.a. &bsim 158 a.a) full-length recombinant protein with GST tag.,MKNK2 (AAH18345 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length recombinant MKNK2.
For Research Use Only
Protein Aliases: G protein-coupled receptor kinase 7; GPRK7; MAP kinase signal-integrating kinase 2; MAP kinase-interacting serine/threonine-protein kinase 2; MAPK signal-integrating kinase 2; MNK2
Gene Aliases: GPRK7; MKNK2; MNK2
UniProt ID: (Human) Q9HBH9
Entrez Gene ID: (Human) 2872
Molecular Function: cytoskeletal protein kinase microtubule family cytoskeletal protein non-motor microtubule binding protein non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.