Novus Biologicals
Manufacturer Code:NBP15812620UL
Catalog # NBP1582620
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to KIF23(kinesin family member 23) The peptide sequence was selected from the middle region of KIF23. Peptide sequence HMQGKLNEKEKMISGQKLEIERLEKKNKTLEYKIEILEKTTTIYEEDKRN. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: kinesin family member 23 kinesin-like 5 (mitotic kinesin-like protein 1) Kinesin-like protein 5 kinesin-like protein KIF23 KNSL5CHO1 mitotic kinesin-like 1 MKLP-1 MKLP1Mitotic kinesin-like protein 1; kinesin-like 5 (mitotic kinesin-like protein 1); Kinesin-like protein 5; Kinesin-like protein KIF23; Mitotic kinesin-like protein 1
Gene Aliases: CHO1; KIF23; KNSL5; MKLP-1; MKLP1
UniProt ID: (Human) Q02241
Entrez Gene ID: (Human) 9493
Molecular Function:
cytoskeletal protein
microtubule binding motor protein
microtubule family cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.