Proteintech
Manufacturer Code:211661AP150UL
Catalog # 50-173-3294
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MKL1 Fusion Protein Ag15240 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: basic, SAP and coiled-coil domain; BSAC KIAA1438 MAL MKL/myocardin like protein 1 MKL1 MRTF A MRTFA MRTF-A; Megakaryoblastic leukemia 1 protein; Megakaryocytic acute leukemia protein; MKL/myocardin-like protein 1; MRTF-A; Myocardin-related transcription factor A
Gene Aliases: BSAC; KIAA1438; MAL; MKL1; MRTF-A; MRTFA
UniProt ID: (Human) Q969V6
Entrez Gene ID: (Human) 57591
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.