Novus Biologicals
Manufacturer Code:NBP299025H
Catalog # NB057413
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human MKKS. |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bardet-Biedl syndrome 6 protein; Bardet-Biedl syndrome 6 protein BBS6MKS HMCS KMS McKusick-Kaufman syndrome McKusick-Kaufman/Bardet-Biedl syndromes putative chaperonin; McKusick-Kaufman/Bardet-Biedl syndromes putative chaperonin
Gene Aliases: BBS6; HMCS; KMS; MKKS; MKS
UniProt ID: (Human) Q9NPJ1
Entrez Gene ID: (Human) 8195
Molecular Function: chaperone chaperonin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.