Invitrogen
Manufacturer Code:PA5102599
Catalog # PIPA5102599
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human MAP2K7 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2 5930412N11Rik c-Jun N-terminal kinase kinase 2 dual specificity mitogen-activated protein kinase kinase 7 EMBL:AAX61178.1} JN JNK kinase 2 JNK-activating kinase 2 JNKK JNKK 2 JNKK2 MAP kinase kinase 7 MAP2K4 map2k7 map2k7 {ECO:0000312 MAPK/ERK kinase 4 MAPK/ERK kinase 7 MAPKK 4 MAPKK 7 Mapkk7 MEK MEK 4 MEK 7 MEK7 mitogen activated protein kinase kinase 7 mitogen-activated protein kinase kinase 7 Mkk7 PRKMK7 protein kinase mitogen activated kinase 7 SAPK kinase 1 SAPK kinase 4 SAPK/ERK kinase 1 SAPKK1 SAPKK-1 SAPKK4 SAPKK-4 SEK1 sek2 SKK4 Stress-activated protein kinase kinase 4; c-Jun N-terminal kinase kinase 2; Dual specificity mitogen-activated protein kinase kinase 7; JNK kinase 2; JNK-activating kinase 2; JNKK 2; MAP kinase kinase 7; map2k7; map2k7 {ECO:0000312|EMBL:AAX61178.1}; MAPK/ERK kinase 7; MAPKK 7; MEK 7; mitogen activated protein kinase kinase 7; protein kinase, mitogen activated, kinase 7; SAPK kinase 4; Stress-activated protein kinase kinase 4
Gene Aliases: JNKK; 2; 5930412N11Rik; JNKK 2; JNKK2; MAP2K7; MAPKK 7; MAPKK7; MEK; MEK 7; MEK7; MKK7; PRKMK7; SAPKK-4; SAPKK4; sek2; SKK4
UniProt ID: (Human) O14733, (Rat) Q4KSH7, (Mouse) Q8CE90
Entrez Gene ID: (Human) 5609, (Rat) 363855, (Mouse) 26400
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.