Novus Biologicals
Manufacturer Code:NB10056561
Catalog # NB10056561
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 21D1418 |
Immunogen | A synthetic peptide mapping to the 300-350 a.a. region of accession number O75030. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: bHLHe32; BHLHE32 bHLHe32MI Class E basic helix-loop-helix protein 32 microphthalmia-associated transcription factor Waardenburg syndrome type 2A WS2A; Class E basic helix-loop-helix protein 32; microphtalmia-associated transcription factor; Microphthalmia-associated transcription factor
Gene Aliases: BHLHE32; CMM8; MI; MITF; WS2; WS2A
UniProt ID: (Human) O75030
Entrez Gene ID: (Human) 4286
Molecular Function:
basic helix-loop-helix transcription factor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.